Water suitable for ion chromatography, Supelco®

Supplier: SUPELCO


M-Clarity™ quality level = MQ100

00612-2.5L 00612-5L
00612-2.5LEA 55 EUR
00612-2.5L 00612-5L
Water suitable for ion chromatography, Supelco®
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66c87043737e047d8bff27c581f18ad666cd31dd&vid=458b88d4a2fabb7b0fe095c236478ebad2f64d4a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR