D-(+)-Glucose ≥99.5% for biotechnology

Supplier: VWR Chemicals

0188-1KGEA 79.5 EUR
0188-1KG 0188-2.5KG 0188-50KG 0188-12KG 0188-500G 0188-5KG
D-(+)-Glucose ≥99.5% for biotechnology
D(+)-Glucose

Sugar used as an energy source in specialised media.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=18aebec8588c01e468167161dd721370b55777b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Arsenic ≤0.00004%
Chloride ≤0.01%
DNase none detected
Heavy Metals (as Pb) ≤0.0005%
Insolubles ≤0.005%
Iron ≤0.0005%
Loss on Drying ≤0.2%
Protease none detected
Purity ≤99.5%
Residue on Ignition ≤0.02%
RNase none detected
Specific Rotation 52.5 – 54 °
Sulphate & Sulphite ≤0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR