Sodium fluoride, BAKER ANALYZED® ACS, J.T.Baker®

Danger

0285.0500EA 92.76 EUR
0285.0500
Sodium fluoride, BAKER ANALYZED® ACS, J.T.Baker®
Sodium fluoride
Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1700
Melting Pt: 993
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1 III
Merck Index: 13,08691
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=5de299c7c1002dab6bcd571d3b9397558a27a5dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Specifications. Meets Reagent Specificatons for testing USP/NF monographs.
Assay min. 99.0 %
Chloride (Cl) max. 0.005 %
Iron (Fe) max. 0.003 %
Potassium (K) max. 0.02 %
Heavy Metals (as Pb) max. 0.002 %
Insoluble Matter max. 0.02 %
Loss on Drying at 150°C max. 0.2 %
Sodium Fluosilicate (Na2SiF6) max. 0.1 %
Sulfate (SO4) max. 0.02 %
Sulfite (SO3) max. 0.005 %
Titrable Acid (meq/g) max. 0.03
Titrable Base (meq/g) max. 0.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR