Ethylene glycol, high purity

Supplier: VWR Chemicals
Warning

0582-1LEA 116 EUR
0582-1L 0582-4L
Ethylene glycol, high purity
Ethylene glycol

Colourless, slightly viscous liquid. Clear with no visible particulates.

Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: 196 – 198 °C (1013 hPa)
Melting Pt: –13 °C
Density: 1.115 g/cm³ (20 °C)
Flash Pt: 116
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=dd4574b881f49679b9aa38891bceb2890214468a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acidity ≤0.002%
Iron ≤0.0005%
Moisture (KF) ≤0.5%
Purity ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR