D-(-)-Mannitol

Supplier: VWR Chemicals

0880-50KGEA 1620 EUR
0880-50KG
D-(-)-Mannitol
D(-)-Mannitol
Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
MDL Number: MFCD00064287
CAS Number: 69-65-8
Merck Index: 13,05769
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=a880987b6e666d8d6d0f15d1877728a17c3a02ad&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR