Hydrofluoric acid 40%, Suprapur® for trace analysis, Supelco®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ100

1.00335.0500 1.00335.2500 1.00335.1000 1.00335.0250
1.00335.0500EA 121 EUR
1.00335.0500 1.00335.2500 1.00335.1000 1.00335.0250
Hydrofluoric acid 40%, Suprapur® for trace analysis, Supelco®
Hydrofluoric acid
Formula: HF
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8 (6.1),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -44 °C
Boiling Point 112 °C
Density 1.13 g/cm3 (20 °C)
pH (H2O, 20 °C) strongly acid
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR