Hydrofluoric acid 38 - 40%, Supelco®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ200

1.00337.1000 1.00337.2500
1.00337.1000EA 82.4 EUR
1.00337.1000 1.00337.2500
Hydrofluoric acid 38 - 40%, Supelco®
Hydrofluoric acid
Formula: HF
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8 (6.1),II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR