Hydriodic acid 57%, Supelco®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ200

1.00341.0250 1.00341.9022
1.00341.0250EA 204 EUR
1.00341.0250 1.00341.9022
Hydriodic acid 57%, Supelco®
Hydriodic acid
Formula: HI
MW: 127.91 g/mol
Density: 1.7 g/cm³ (20 °C)
MDL Number: MFCD00011347
CAS Number: 10034-85-2
EINECS: 233-109-9
UN: 1787
ADR: 8 II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -50 °C (External MSDS)
Boiling Point 127 °C (1013 hPa)
Density 1.70 g/cm3 (20 °C)
pH 1 (13 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR