4-(Indol-3-yl)butyric acid, Millipore®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ100

1.00354.0005 1.00354.0025 1.00354.0100 1.00354.0500
1.00354.0005EA 59.1 EUR
1.00354.0005 1.00354.0025 1.00354.0100 1.00354.0500
4-(Indol-3-yl)butyric acid, Millipore®
4-(Indol-3-yl)butyric acid
Formula: C₁₂H₁₃NO₂
MW: 203.24 g/mol
Melting Pt: 121…126 °C
Density: 1.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005664
CAS Number: 133-32-4
EINECS: 205-101-5
UN: 2811
ADR: 6.1 III
Merck Index: 13,04987

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR