Molybdenum(VI) oxide analytical reagent, Supelco®

Supplier: MERCK MILLIPORE
Warning


M-Clarity™ quality level = MQ300

1.00403.0100 1.00403.0500
1.00403.0100EA 178 EUR
1.00403.0100 1.00403.0500
Molybdenum(VI) oxide analytical reagent, Supelco®
Molybdenum (VI) oxide
Formula: MoO₃
MW: 143.94 g/mol
Boiling Pt: 1155
Melting Pt: 795
Density: 4.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
UN: 3288
Merck Index: 13,06261

Order Now

Specification Test Results

Solubility 0.5 g/l (20 °C)
Melting Point 795 °C
Molar Mass 143.94 g/mol
Bulk Density 1200 kg/m3
Boiling Point 1155 °C
Density 4.70 g/cm3 (26 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR