4-(Dimethylamino)benzaldehyde Reag. Ph. Eur. analytical reagent, Supelco®

Supplier: MERCK MILLIPORE
Warning


M-Clarity™ quality level = MQ100

1.03058.0025 1.03058.0100 1.03058.1000
1.03058.0025EA 99.6 EUR
1.03058.0025 1.03058.0100 1.03058.1000
4-(Dimethylamino)benzaldehyde Reag. Ph. Eur. analytical reagent, Supelco®
4-(Dimethylamino)benzaldehyde
Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: 176…177 °C (1013 hPa)
Melting Pt: 72
Density: 1.0254 g/cm³ (99.9 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
EINECS: 202-819-0
UN: 3077
ADR: 9 III
Merck Index: 13,03257
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=a978f8cb2be9fb6dfa0aa34ce0038883c864820b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.3 g/l (20 °C)
Melting Point 72 - 75 °C
Molar Mass 149.19 g/mol
Bulk Density 350 kg/m3
Boiling Point 176 - 177 °C (23 hPa)
Flash Point 100 °C
Ignition Point 445 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR