Dimethyl sulphoxide-[D₆] (99.9% D) with TMS (0.1 vol.%), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Supplier: MERCK MILLIPORE


M-Clarity™ quality level = MQ200

1.03587.0100 1.03587.0006
1.03587.0100EA 541 EUR
1.03587.0100 1.03587.0006
Dimethyl sulphoxide-[D₆] (99.9% D) with TMS (0.1 vol.%), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
MW: 84.09 g/mol
Boiling Pt: 189
Melting Pt: 20.2
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 82 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Deuteration degree ≥ 99.9 %
Water (K. Fischer, H₂O + D₂O) ≤ 0.030 %
Tetramethylsilane (NMR-spectrum) 0.001
Water (NMR, H₂O) ≤ 0.020 %
Performance test (NMR-spectrum) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR