n-Hexane ≥99.0% (by GC), Suprasolv® hypergrade for organic trace analysis, Supelco®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ100

1.04369.1000 1.04369.2500 1.04369.9010
1.04369.1000EA 101 EUR
1.04369.1000 1.04369.2500 1.04369.9010
n-Hexane ≥99.0% (by GC), Suprasolv® hypergrade for organic trace analysis, Supelco®
n-Hexane

Suprasolv® hypergrade is a unique and universal solvent quality which can be employed for all three GC detection methods (ECD, FID and MS). Thanks to the large retention time range, even low-boiling substances can be reliably analysed.

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: –94.3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c939cebcd37af6287105861069a46c7635aa4af8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥99.0%
Sum of Hexane Isomers+Methylcyclopentane (GC) ≥99.9%
Identity (IR) Conforms
Evaporation Residue ≤3.0 mg/l
Water ≤0.005%
Color ≤10 Hazen
GC/ECD (Retention Range 1,2,4-tri- Chlorobenzene to Decachlorobiphenyle, Individual Signals (Lindane Standard)) ≤2 pg/ml
GC/ECD (Retention range Dichloromethane to 1,2,4-Trichlorobenzene Individual Signals (Tetrachloromethane)) ≤1 ng/ml
GC/FID (Retention Range n-Undecane to n-Tetracontane, Individual Signals (n-Tetradecane Standard)) ≤2 ng/ml
GC/MSD (Retention Range n-Undecane to n-Tetracontane, Scaning Area 30 - 600 amu, Individual Signals (n- Tetradecane Standard) ≤2 ng/ml
Recommended for Analysis of Polychlorinated Dibenzodioxins and Polychlorinated Dibenzofurans (PCDD/F).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR