Sodium hydroxide 30% in aqueous solution, Suprapur® for trace analysis, Supelco®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ100

1.05589.0250 1.05589.1000 1.05589.2500 1.05589.0500
1.05589.0250EA 444 EUR
1.05589.0250 1.05589.1000 1.05589.2500 1.05589.0500
Sodium hydroxide 30% in aqueous solution, Suprapur® for trace analysis, Supelco®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.33 g/cm3 (20 °C)
pH 14 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR