Manganese(II) sulphate monohydrate, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ300

1.05941.0250 1.05941.9025
1.05941.0250EA 69.3 EUR
1.05941.0250 1.05941.9025
Manganese(II) sulphate monohydrate, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
Manganese (II) sulphate monohydrate
Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700
Density: 2.95 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9 III
Merck Index: 13,05763

Order Now

Specification Test Results

Solubility 762 g/l (20 °C)
Melting Point 700 °C (anhydrous substance)
Molar Mass 169.02 g/mol
Bulk Density 1000 - 1200 kg/m3
Density 2.95 g/cm3 (20 °C)
pH 3.0 - 3.5 (50 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous hazardous material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR