Paraffin, liquid_highly NF, JP, Ph. Eur., BP, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS
Danger

1.07174.1000 1.07174.2500 1.07174.9026 1.07174.9180
1.07174.1000EA 108 EUR
1.07174.1000 1.07174.2500 1.07174.9026 1.07174.9180
Paraffin, liquid_highly NF, JP, Ph. Eur., BP, SAFC®
Paraffin

Residues of metal catalysts or metal reagents acc. to EMEA/C HMP/SWP/4446/2000 are not likely to be present.
Corresponds to Ph Eur,BP,NF,JP

Density: 0.85 g/cm³ (20 °C)
Flash Pt: >180 °C
Storage Temperature: Ambient
CAS Number: 8002-74-2
EINECS: 232-455-8

Order Now

Specification Test Results

Identity passes test
Purity passes test
Acidity or alkalinity passes test
Density (d 20 °C/20 °C) 0.830 - 0.870
Dynamic viscosity (20 °C) 25.0 - 80.0 mPa∙s
Kinematic Viscosity (40 °C) ≤ 33.5 mm²/s
Arsenic (As) ≤ 2 ppm
Heavy metals (as Pb) ≤ 10 ppm
Residual solvents (Ph.Eur./ICH) excluded by manufactoring process
Aromatic, polycyclic Hydrocarbons passes test
Solid Paraffines passes test
Readily carbonisable substance passes test
Sulphur compounds passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR