Cleansers, liquid concentrates and powders, for use with washing machines, AP series, Extran®, Supelco®
Supplier: Merck
1.07558.2000 1.07558.9025 1.07559.2500 1.07559.9010 1.07559.9025 1.07561.2500 1.07561.9010 1.07561.9025 1.07563.2000 1.07563.9010 1.07563.9025 1.07570.2000 1.07570.9025 1.07558.9010 1.40006.2500 1.40006.9010 1.40006.9025 1.40006.5000 1.40118.2500 1.40118.5000 1.40118.9010 1.40118.9025 1.07559.5001 1.07561.5001 1.40006.5001 1.40118.5001
1.07558.2000EA 215 EUR
1.07558.2000 1.07558.9025 1.07559.2500 1.07559.9010 1.07559.9025 1.07561.2500 1.07561.9010 1.07561.9025 1.07563.2000 1.07563.9010 1.07563.9025 1.07570.2000 1.07570.9025 1.07558.9010 1.40006.2500 1.40006.9010 1.40006.9025 1.40006.5000 1.40118.2500 1.40118.5000 1.40118.9010 1.40118.9025 1.07559.5001 1.07561.5001 1.40006.5001 1.40118.5001
Cleansers, liquid concentrates and powders, for use with washing machines, AP series, Extran®, Supelco®
Cleaning Agents Detergents and Surface Cleansers
A range of cleansers, for machine washing and rinsing, that have been developed to meet the cleaning requirements for glassware and small equipment in the laboratory.
  • Prevents carry-over of residues to the next analysis
  • Chlorine-free
These products have been developed with leading instrument manufacturers for their suitability for washing machines. Extran® products have been tested extensively for their effectiveness and low degree of foaming. It is advisable to neutralise alkaline cleansers with an acid rinse subsequent to the cleaning process.More Product Information