Tetrahydrofuran, LiChrosolv® for liquid chromatography, Supelco®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ100

1.08101.1000 1.08101.2500 1.08101.9010 1.08101.9030 1.08101.9015 1.08101.4000
1.08101.1000EA 215 EUR
1.08101.1000 1.08101.2500 1.08101.9010 1.08101.9030 1.08101.9015 1.08101.4000
Tetrahydrofuran, LiChrosolv® for liquid chromatography, Supelco®
Tetrahydrofuran

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in 'gradient grade' as well as 'isocratic grade'. This minimises the gradient effect of the solvent involved.

Formula: C₄H₈O
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108,5 °C
Density: 0,888 g/cm³ (20 °C)
Flash Pt: –21,5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=11aea9af60d6f59ae6301982bc1178f0ad862d59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -108.5 °C
Molar Mass 72.11 g/mol
Boiling Point 65 - 66 °C (1013 hPa)
Vapor Pressure 173 hPa (20 °C)
Flash Point -20 °C
Refractive Index
Explosion Limit 1.5 - 12.4 %(V)
Density 0.89 g/cm3 (20 °C)
pH 7 - 8 (200 g/l, H2O, 20 °C)
Ignition Point 215 °C
Viscosity kinematic
Synonyms THF, Tetramethylene oxide, Oxolane, Butylene oxide
Hill Formula C4H8O
Chemical Formula

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR