Tetrahydrofuran, Uvasol® for spectroscopy, Supelco®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ100

1.08110.0500 1.08110.2500
1.08110.0500EA 163 EUR
1.08110.0500 1.08110.2500
Tetrahydrofuran, Uvasol® for spectroscopy, Supelco®
Tetrahydrofuran
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=11aea9af60d6f59ae6301982bc1178f0ad862d59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -108.5 °C
Molar Mass 72.11 g/mol
Boiling Point 65 - 66 °C (1013 hPa)
Vapor Pressure 173 hPa (20 °C)
Flash Point -20 °C
Explosion Limit 1.5 - 12.4 %(V)
Density 0.89 g/cm³ (20 °C)
pH 7 - 8 (200 g/L, H₂O, 20 °C)
Ignition Point 215 °C
Saturation Concentration 557 g/m³ (20 °C) Air
Evaporation Number 2.3
Hazard Class 3
Hazard Class Description Flammable liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR