Vanillin, EMPROVE® ESSENTIAL BP, Ph. Eur., NF suitable for use as excipient, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS
Warning


MQ100

1.08510.9025 1.08510.1000
1.08510.9025EA 0 EUR
1.08510.9025 1.08510.1000
Vanillin, EMPROVE® ESSENTIAL BP, Ph. Eur., NF suitable for use as excipient, SAFC®
Vanillin
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 285
Melting Pt: ∼82 °C
Density: 1.056 g/cm³ (20 °C)
Flash Pt: 153
Storage Temperature: Ambient
MDL Number: MFCD00006942
CAS Number: 121-33-5
EINECS: 204-465-2
Merck Index: 13,09998
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd6583c5aea50cd090e103650a05eab491b4d38e&vid=fd6583c5aea50cd090e103650a05eab491b4d38e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR