Zinc ≥99.9%, coarse powder, EMSURE® Reag. Ph. Eur. ∼ 0.3- 1.5 mm (14-50 mesh ASTM) for analysis, suitable for filling of reductors, Supelco®

Supplier: MERCK MILLIPORE
Warning


M-Clarity™ quality level = MQ300

1.08756.0250 1.08756.1000
1.08756.0250EA 353 EUR
1.08756.0250 1.08756.1000
Zinc ≥99.9%, coarse powder, EMSURE® Reag. Ph. Eur. ∼ 0.3- 1.5 mm (14-50 mesh ASTM) for analysis, suitable for filling of reductors, Supelco®
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 419 °C
Density: 7.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 3077
ADR: 9 III
Merck Index: 12,10255

Order Now

Specification Test Results

Assay (complexometric) ≥ 99.9 %
As (Arsenic) ≤ 0.00001 %
Cd (Cadmium) ≤ 0.002 %
Cu (Copper) ≤ 0.001 %
Fe (Iron) ≤ 0.02 %
Pb (Lead) ≤ 0.005 %
Sn (Tin) ≤ 0.001 %
Corresponds to Reag. Ph Eur


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR