Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in 2-propanol/methanol, Titripur® Reag. USP, Reag. Ph. Eur. for titration in non-aqueous media, Supelco®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ100

1.09162.0500 1.09162.1000 1.09162.1003
1.09162.0500EA 135 EUR
1.09162.0500 1.09162.1000 1.09162.1003
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in 2-propanol/methanol, Titripur® Reag. USP, Reag. Ph. Eur. for titration in non-aqueous media, Supelco®
Tetrabutylammonium hydroxide

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Melting Pt: 29.85
Density: 0.968
Storage Temperature: Ambient
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 1987
ADR: 3 II

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.0994 - 0.1006
Measurement uncertainty +/- 0.0004
Traceability NIST SRM
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR