Lanthanum(III) oxide, Supelco®

Supplier: MERCK MILLIPORE


M-Clarity™ quality level = MQ200

1.12220.0100 1.12220.0500
1.12220.0100EA 195 EUR
1.12220.0100 1.12220.0500
Lanthanum(III) oxide, Supelco®
Lanthanum (III) oxide
Formula: La₂O₃
MW: 325.81 g/mol
Boiling Pt: 4200
Melting Pt: 2315
Density: 6.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011071
CAS Number: 1312-81-8
EINECS: 215-200-5

Order Now

Specification Test Results

Melting Point 2315 °C
Molar Mass 325.82 g/mol
Bulk Density 1300 kg/m3
Boiling Point 4200 °C
Density 6.51 g/cm3 (20 °C)
pH 9.0 (50 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR