N-Acetyl-L-cysteine, Millipore®

Supplier: MERCK MILLIPORE
Warning


M-Clarity™ quality level = MQ100

1.12422.0025 1.12422.0100 1.12422.1000 1.12422.9010
1.12422.0025EA 46.9 EUR
1.12422.0025 1.12422.0100 1.12422.1000 1.12422.9010
N-Acetyl-L-cysteine, Millipore®
N-Acetyl-L-cysteine
Formula: C₅H₉NO₃S
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 106…108 °C
Density: 1,48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004880
CAS Number: 616-91-1
EINECS: 210-498-3
Merck Index: 13,00090

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR