Acetonitrile, LiChrosolv®, Isocratic Grade for liquid chromatography, Supelco®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ100

1.14291.1000 1.14291.2500 1.14291.4000 1.14291.9010 1.14291.9030 1.14291.9185 1.14291.9910
1.14291.1000EA 199 EUR
1.14291.1000 1.14291.2500 1.14291.4000 1.14291.9010 1.14291.9030 1.14291.9185 1.14291.9910
Acetonitrile, LiChrosolv®, Isocratic Grade for liquid chromatography, Supelco®
Acetonitrile

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3
REACH: 01-2119471307-38
Merck Index: 13,00071
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b965ef40b6e61dd29b33363d25c3df2038fea764&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR