Potassium hexachloroplatinate(IV) ≥99%, Supelco®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ100

1.19238.0001 1.19238.0005
1.19238.0001EA 212 EUR
1.19238.0001 1.19238.0005
Potassium hexachloroplatinate(IV) ≥99%, Supelco®
Potassium hexachloroplatinate(IV)
Formula: K₂PtCl₆
MW: 485.99 g/mol
Melting Pt: 250
Density: 3.344 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011389
CAS Number: 16921-30-5
EINECS: 240-979-3
UN: 2923
ADR: 8 (6.1)
Merck Index: 13,07720

Order Now

Specification Test Results

Identity (elemental analysis) passes test
Assay (calculated of Pt) 99.0 - 101.0 %
Pt (Platinum) 39.7 - 40.5 %
Al (Aluminium) ≤ 10 ppm
Ca (Calcium) ≤ 50 ppm
Cd (Cadmium) ≤ 10 ppm
Co (Cobalt) ≤ 10 ppm
Cr (Chromium) ≤ 10 ppm
Cu (Copper) ≤ 50 ppm
Fe (Iron) ≤ 50 ppm
Hg (Mercury) ≤ 100 ppm
Ir (Iridium) ≤ 100 ppm
Mg (Magnesium) ≤ 50 ppm
Mn (Manganese) ≤ 10 ppm
Ni (Nickel) ≤ 10 ppm
Os (Osmium) ≤ 100 ppm
Pb (Lead) ≤ 50 ppm
Pd (Palladium) ≤ 100 ppm
Rh (Rhodium) ≤ 100 ppm
Ru (Ruthenium) ≤ 100 ppm
Zn (Zinc) ≤ 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR