Acetic acid ≥80%, EMPROVE® EXPERT for biopharmaceutical production

Supplier: MERCK PRODUCTION CHEMICALS
Danger

1.37072.1000
1.37072.1000EA 83.5 EUR
1.37072.1000
Acetic acid ≥80%, EMPROVE® EXPERT for biopharmaceutical production
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2790
ADR: 8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=031896546f0eb78526a541f0af4636e3991e9f8e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR