Face shields, Supervizor™
Supplier: HONEYWELL SAFETY
1002295 1002297 1002309 1002302 1002304 1002305 1002310 1002312 1002329 1002330 1004583 1004584 1004587 1002308
1002295.EA 12 EUR
1002295. 1002297. 1002309. 1002302. 1002304. 1002305. 1002310. 1002312. 1002329. 1002330. 1004583. 1004584. 1004587. DALL1002308
Face shields, Supervizor™
Face Shields
Robust, heavy-duty, helmet mounted face shield range with hearing protection compatibility. The products provide protection from abrasion, chemical hazards, gas, vapors and impact - they are therefore ideal for chemical applications or forestry.
  • Secure visor fixing methods allow easy visor replacement and maximum protection, allowing full unrestricted head movement
  • Items are compatible with a wide range of helmet designs and helmet mounted earmuffs (including Bilsom communication earmuffs)
  • Browguard with ratchet headband provides additional safety and
More Product Information