Face shields, Clearways
Supplier: HONEYWELL SAFETY
1002360 1002353 1002354 1002359 1002369 1002370 1002366
1002360.EA 13.7 EUR
1002360. 1002353. 1002354. 1002359. 1002369. 1002370. 111-1360
Face shields, Clearways
Face Shields
This combination offers both strength and durability. The visors are suitable for different kinds of applications.
  • Lightweight - comfortable enough to wear all day
  • Integral visor fitting ensures extra protection against chemical seepage
  • Economical replaceable visor available in acetate or PC (PC: High impact; acetate: Chemical resistant)
Weight: 160 g
Visor: 200 mm
EN 166
Certifications: EN 166
Ordering information: Visor, brow guard, and chin guard must be ordered separately. See Accessories tab for full component listings. Holder mustMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable