Sodium dihydrogen phosphate monohydrate 99.0-102.0%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals

102454REA 93.2 EUR
102454R 102455S
Sodium dihydrogen phosphate monohydrate 99.0-102.0%, AnalaR NORMAPUR® analytical reagent
Sodium dihydrogen phosphate monohydrate
Formula: NaH₂PO₄·H₂O
Melting Pt: 100 °C
Density: 2,04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 13,08734
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=5597299de719468db5a68462cc750045659b46af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 - 102.0 %
Solution in water Passes test
pH (5 %) 4.1 - 4.5
Cl (Chloride) Max. 0.0005 %
SO4 (Sulphate) Max. 0.005 %
Total N (Nitrogen) Max. 0.001 %
Heavy metals (as Pb) Max. 0.0005 %
As (Arsenic) Max. 0.00005 %
Ca (Calcium) Max. 0.005 %
Cu (Copper) Max. 0.0002 %
Fe (Iron) Max. 0.0005 %
K (Potassium) Max. 0.005 %
Pb (Lead) Max. 0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR