Mixed cellulose ester membranes, non-sterile, 0,45 µm, Whatman™
Supplier: GE Healthcare - Whatman
7141-004 10401606 10401612 10401614 10401618 10401621 10401626 10401631 10406512 10406812 10406814 10409414 10409714 10401664 7141-204
7141004.EA 240 EUR
7141004. 10401606. 10401612. 10401614. 10401618. 10401621. 10401626. 10401631. 10406512. 10406812. 10406814. 10409414. 10409714. 514-0944 7141204.
Mixed cellulose ester membranes, non-sterile, 0,45 µm, Whatman™
Filters Membrane Filters
Mixed cellulose ester, WME range, circles, gridded. Whatman mixed cellulose ester membranes are composed of cellulose acetate and cellulose nitrate. These membranes are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
  • Sterile options available for critical applications
  • Excellent contrast for easier particle detection
  • Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
  • Black plain and black gridded membranes have a mix of cellulose nitrate and cellulose acetate
  • The membrane offers a high
More Product Information