Amersham™ Protran® Premium Western Blotting Membranes, Nitrocellulose
Supplier: Cytiva
10600034 10600047 10600048 10600003 10600004 10600008 10600009 10600013 10600014 10600096 10600117 10600118 10600065 10600079 10600080
10600034.EA 435 EUR
10600034. 10600047. 10600048. 10600003. 10600004. 10600008. 10600009. 10600013. 10600014. 10600096. 10600117. 10600118. 10600065. 10600079. 10600080.
Amersham™ Protran® Premium Western Blotting Membranes, Nitrocellulose
Transfer Membranes
Amersham™ Protran® Premium nitrocellulose blotting membranes provide excellent sensitivity, resolution and low background in fluorescence Western blotting detection applications.
  • Available in popular pre-cut sizes
  • High binding capacity
  • Protran® Premium 0,2 NC offers low auto-fluorescence across a wide range of excitation/emission wavelengths, making the membrane suitable for fluorescence-based immunodetection and multiplexing
  • Protran® Premium 0,45 NC is an unsupported, 100% pure nitrocellulose membrane suitable for proteins of a wide range of molecular
More Product Information