Earmuffs, Peltor™ Optime™ III
Supplier: 3M
H540A H540B H540P3E
111-2202EA 70 EUR
111-2202 111-1083 111-1085
Earmuffs, Peltor™ Optime™ III
Earmuffs Noise Blocking Earmuffs
These earmuffs have been developed for use in extremely high noise environments, particularly those dominated by low frequency sounds. The protection is based on a technology with a double casing, minimising resonance in the holder casing resulting in maximum high frequency attenuation.
  • Broad sealing rings filled with soft plastic foam for optimum fit and low contact pressure
  • Good speech intelligibility
  • Easy to replace cushions and inserts help keep them hygienically clean
  • Helmet-mounted version is designed to fit a wide range of industrial safety helmets
More Product Information