Earmuffs, Peltor™ X series
Supplier: 3M
X5A X5P3E X1P3E X2A X2P3E X3A X3P3E X4A X4P3E X1A
111-1139EA 97.6 EUR
111-1139 111-1141 111-1132 111-1133 111-1134 111-1135 111-1136 111-1137 111-1138 111-1131
Earmuffs, Peltor™ X series
Earmuffs Noise Blocking Earmuffs
These earmuffs combine excellent protection with ultimate comfort. They are available as headband and helmet-mounted versions, offering a wide range of attenuation levels that meet the needs of the majority of industrial applications.
  • Soft, wide cushions reduce pressure around the ears and improve comfort and wearability
  • Slimline, dual-moulded cups
  • Large space inside cups reduces moisture and heat build-up
  • Electrically-insulated wire headband
  • Colour coded cups: Easy to identify level of attenuation
X1: Ultra-slim earmuffs for low noise environments.
X2:More Product Information