Disposable particle filtering respirators, FFP2/FFP3, Medical Mask
Supplier: MOLDEX
3150 3100 3250 3200
111-1146EA 41.6 EUR
111-1146 111-1145 111-1148 111-1147
Disposable particle filtering respirators, FFP2/FFP3, Medical Mask
Respirators Disposable Respirators Particulate Respirators
These air masks, without exhalation valves, are medical masks and FFP masks in one. They offer protection for both the wearer and their surroundings. Due to the Moldex pleated filter technology, the masks combine the filtering performance of an FFP mask with the low breathing resistance of a conventional medical mask.
  • Combined FFP mask and medical mask
  • Significantly reduced breathing resistance, high filter capacity, longer usage time
  • Available in two sizes
  • DuraMesh® - strong and durable structure, keeps the mask in shape
  • ActivForm® - no nose clip needed
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable