Maintenance-free half mask respirators, 4000 series
Supplier: 3M
4251PLUS 4255PLUS 4277PLUS 4279PLUS
111-2443EA 50.8 EUR
111-2443 111-2444 111-2588 111-2445
Maintenance-free half mask respirators, 4000 series
Respirators Reusable Respirators Half Face Respirators
4000 series maintenance-free half masks are simple and ready to use respirators with an integrated cartridge and filters. They provide protection against a range of organic gases and vapours, also combination particulate hazards.
  • Newly enhanced valve assembly reduces exhalation breathing resistance for improved breathing comfort
  • Lightweight, well balanced, low profile design
  • Integrated cartridge and filters are ready to use and maintenance-free
  • Adjustable head cradle and easy to fasten neck straps
  • Centrally positioned exhalation valve reduces heat and
More Product Information