Chemical resistant gloves, PowerCoat® 948-31 Nitraf
Supplier: HONEYWELL SAFETY
2094831/8 2094831/9 2094831/10
112-0943EA 0 EUR
112-0943 112-1000 112-1012
Chemical resistant gloves, PowerCoat® 948-31 Nitraf
Gloves
These nitrile dipped gloves with cotton flock lined interior are particularly suitable for handling oils, fats and hydrocarbon derivatives.
  • Nitrile ensures good chemical properties and excellent mechanical characteristics (abrasion)
  • Cotton flock interior ensures maximum user comfort
  • Rough surface finish improves grip in oily or damp environments
  • Cuff protects a large part of the forearm
Ideal for food preparation (greasy products), production of paints and varnishes. Please note: Not suitable for contact with ketone-based solvents.
Thickness: 0,51More Product Information