VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
112-4193EA 29.5 EUR
112-4193 112-4197 112-4194 112-4195 112-4196
VWR® Nitrile Extra Light, Nitrile Gloves
Gloves
These thin, ambidextrous gloves are suitable for use in medical applications, food processing and for general lab requirements in non hazardous areas.
  • Non sterile, powder-free, textured fingertips
  • Very thin: Provide excellent elasticity for good wearer comfort
  • Free from phthalates/softeners and allergenic latex proteins; particularly skin friendly for users with latex allergy
  • Beaded cuffs for easier donning
  • Low thickness and textured fingertips for excellent grip
Certifications: EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS, EN 455 1-4; ASTM D6319, ASTM F1More Product Information