Cleanroom gloves, nitrile, sterile, BioClean Emerald™
Supplier: NITRITEX
BENS8.0 BENS8.5 BENS10.0 BENS6.0 BENS6.5 BENS7.0 BENS7.5 BENS9.0
112-4959EA 616 EUR
112-4959 112-6364 112-4954 112-4955 112-4956 112-4957 112-4958 112-5111
Cleanroom gloves, nitrile, sterile, BioClean Emerald™
Gloves
Hand-specific chemotherapy gloves with beaded cuffs. Suitable for double-donning and for handling cytotoxic drugs.
  • Sterile, powder-free, textured finish
  • Latex-free to eliminate Type 1 latex allergies
  • Hypoallergenic
  • Nitrile is resistant to a wide range of chemicals
  • Each size carries a unique bar code on the carton (ITF-14), which allows automatic data entry into stock control systems
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
EN 455, EN 374-2; Cat. III; AQL 1,5
Certifications: EN 455, EN ISO 374-1 Type B, EN ISO 374-5 VIRUS, EN 421,More Product Information