VWR®, Chemical Resistant Gloves, Nitrile, Flock-Lined
Supplier: VWR Collection
112-5656EA 47 EUR
112-5656 112-5652 112-5653 112-5654 112-5655
VWR®, Chemical Resistant Gloves, Nitrile, Flock-Lined
Gloves
Gloves are made from a specially formulated acrylonitrile butadiene compound that provides outstanding protection against a wide range of chemicals, including strong detergents, greases, oils, most solvents and acids.
  • Superior resistance to abrasion, cut and puncture when compared to natural rubber gloves
  • Higher durability and longer wear than natural rubber gloves
  • Do not contain latex proteins
  • Patterned palm and fingers provide excellent grip in both wet and dry conditions
  • Cotton flock lining offers added comfort and better absorption of perspiration