VWR®, Precision Grip Gloves, PU Coating
Supplier: VWR Collection
112-5697EA 14.4 EUR
112-5697 112-5698 112-5699 112-5726 112-5727 112-5728
VWR®, Precision Grip Gloves, PU Coating
Gloves
VWR® precision grip gloves with polyurethane coating provide basic cut protection and protection against mechanical risks.
  • ANSI A1 cut protection (Level A)
  • Nylon composition
  • Polyurethane (PU) coating on palm and fingertips
  • Black knitted glove with black coating
Ideal for handling small components, or situations that require dexterity in a dry and/or dirty environment. Also great for package handling, picking and packing.
Certifications: Meets requirements for ANSI A1 Cut Resistance. EN 388:2016 (4131X)
Certified by OKEO-TEX meeting Standard 100More Product Information