Chemical resistant overalls, Tyvek® 500 Xpert, models CHF5 / CHF5a
Supplier: DuPont
TYCHF5SGR2X010 TYCHF5SBUSM010 TYCHF5SGRSM010 TYCHF5SGRMD010 TYCHF5SGRLG010 TYCHF5SGRXL010 TYCHF5SBUMD010 TYCHF5SBULG010 TYCHF5SBUXL010 TYCHF5SBU2X010 TYCHF5SWHLG01P TYCHF5SWHSM01P TYCHF5SWHMD01P TYCHF5SWHXL01P TYCHF5SWH2X01P TYCHF5SWH3X01P TYCHF5SWH2X01B TYCHF5SWHXL01B TYCHF5SWHSM01B TYCHF5SWH3X01B TYCHF5SWHMD01B TYCHF5SWHLG01B TYCHF5SBU3X010 TYCHF5SGR3X010 TYCHF5SWH4X01P TYCHF5SWH5X01P TYCHF5SWH6X01P TYCHF5SWH7X01P
113-3868EA 422 EUR
113-3868 113-3869 113-3864 113-3865 113-3866 113-3867 113-3870 113-3871 113-3872 113-3873 113-6062 113-6060 113-6061 113-6063 113-6064 113-6065 113-8343 113-8342 113-8339 113-8344 113-8340 113-8341 113-9682 113-9683 DUPOTYCHF5SWH4X01P DUPOTYCHF5SWH5X01P DUPOTYCHF5SWH6X01P DUPOTYCHF5SWH7X01P
Chemical resistant overalls, Tyvek® 500 Xpert, models CHF5 / CHF5a
Disposable Garments
These Tyvek® overalls are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
  • Ultra-low linting fabric is robust yet lightweight (<180 g per garment)
  • 3-piece hood for optimal fit to head and face when turning
  • Large, 'easy to grasp' zip pull
  • Elasticated face, wrists and ankles as well as glued-in waist elastic
More Product Information