Surgical masks, tie-on, Type II
Supplier: 3M MEDICAL
M1810F
113-9994EA 70.1 EUR
113-9994
Surgical masks, tie-on, Type II
Masks Surgical Masks
These flat pleated surgical masks with adjustable tie provide comfort, breathability and maximum protection.
  • Provides 98% bacterial filtration efficiency
Certifications: EN 14683:2005
Packaging: Pack of 100 per box and six boxes per case.