Insole, semi-orthopaedic, sole mid
Supplier: ELTEN
204081/36 204081/37 204081/38 204081/39 204081/40 204081/41 204081/42 204081/43 204081/44 204081/45 204081/46 204081/47 204081/48
114-7580EA 8.1 EUR
114-7580 114-7581 114-7582 114-7583 114-7584 114-7585 114-7586 114-7587 114-7588 114-7589 114-7590 114-7591 114-7592
Insole, semi-orthopaedic, sole mid
Shoe Insoles
Semi-orthopaedic inlay sole provides individual cushioning and weight distribution, relieving pain and pressure points.
  • Anatomically shaped insole supports the natural heel-to-toe movement of the foot
  • Excellent cushioning properties provide relief for joints and back
  • Skin friendly materials improve the foot climate
Suitable for ESD safety shoes (electrical resistance of 10⁵ to 10⁷ Ohm).
Colour: Black/blue
Certifications: DGUV 112-191; EN ISO 20345; EN ISO 20347
Order Now
SPECIFICATIONS
Environmentally Preferable