ESD safety ankle boots, lace-up, Miss L10, Sensation Lady Mid
Supplier: ELTEN
74206/35 74206/36 74206/37 74206/38 74206/39 74206/40 74206/41 74206/42
114-8781EA 83.9 EUR
114-8781 114-8782 114-8783 114-8784 114-8785 114-8786 114-8787 114-8788
ESD safety ankle boots, lace-up, Miss L10, Sensation Lady Mid
Shoes and Boots
Casual, vintage style ESD ankle boot for ladies made from hydrophobised textile material in 'denim look', and hydrophobised nubuck leather, a padded tongue plus a low profile dual-density TPU/PU sole (Miss L10).
  • Bioactive fabric lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro Lady), grey
  • Soft fleece ESD insole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected. The shoe is tailored to the ergonomics of female feet.
Colour: Black/grey
Certifications: EN 61340-5-1 (More Product Information