ESD safety ankle boots, lace-up, L10, Sensation Up Mid
Supplier: ELTEN
76201/39 76201/40 76201/41 76201/42 76201/43 76201/44 76201/45 76201/46 76201/47
114-8823EA 84.9 EUR
114-8823 114-8824 114-8825 114-8826 114-8827 114-8828 114-8829 114-8830 114-8831
ESD safety ankle boots, lace-up, L10, Sensation Up Mid
Shoes and Boots
Vintage and casual style, environmental friendly manufactured ESD safety shoe made from made from hydrophobised textile material in jeans look and hydrophobised nubuck leather, a padded tongue and a low profile dual-density TPU/PU sole L10.
  • Fabric lining Bioactive
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO grey
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
More Product Information