Cleanroom wipes, AlphaWipe®
Supplier: Texwipe
TX1004 TX1013 TX1009B TX1009
115-0496EA 239 EUR
115-0496 115-0454 115-1769 115-1949
Cleanroom wipes, AlphaWipe®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These premium heavy-weight all-purpose wipes are made from 100% continuous filament polyester in a double-knit, no run, interlocked pattern. Soft and non abrasive, they are ideal for critical environments where contamination control is essential.
  • Ultra-low particulate and fibre levels
  • Excellent for sensitive surfaces
  • Superior strength guards against abrasion under rigorous use
Suitable for use in Class 10 to 100000, ISO 4 to 8 cleanrooms.
Colour: White.
Caution: Data accurate at time of publishing, latest test results and test methods can beMore Product Information