Cleanroom wipes, multipurpose, Amplitude™ Sigma™
Supplier: CONTEC
AMSI0001 AMSI0003 AMSI0002
115-0573EA 41.2 EUR
115-0573 115-0609 CONTAMSI0002
Cleanroom wipes, multipurpose, Amplitude™ Sigma™
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Non woven cellulose/polyester hydroentangled wipes made from 55% cellulose and 45% polyester, highly sorbent with excellent durability and good cleanliness.
  • Strong and durable, wet or dry
  • Low particle and extractables levels
  • Also available pre-saturated as PROSAT® and SATWipes® wipes
Ideal for non abrasive applications, general purpose cleaning or spillage pick up in biotech, pharmaceutical, semiconductor, electronics, medical device and printed circuit board industries.
Suitable for use in Class 1000, ISO 6 environments.
Colour: White