VWR® B'Wipes 7000, Cleanroom wipes
Supplier: VWR Collection
115-1824EA 78 EUR
115-1824 115-1829
VWR® B'Wipes 7000, Cleanroom wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Microfibre, sealed-edge wipe for use in ISO 3 cleanrooms.
  • Microfibre polyester/nylon wipe, with sealed edges (165 g/m²)
  • Laundered, with sealed edges, for excellent cleanliness
  • Very fine microfibre filaments, densely knitted, greatly increase the quantity of contaminants picked up by the wipe, even when dry
  • Reduces the risk of scratching sensitive surfaces, such as optics
  • Ideal for cleaning sensitive surfaces, or when wet wiping is not an option