Cleanroom wipes, pre-saturated, sterile, PROSAT® Sterile™ Low Endotoxin
Supplier: CONTEC
PSWE0001-BPR PSWE0002-BPR PSWE0003-BPR
115-2821EA 1730 EUR
115-2821 115-2822 115-2823
Cleanroom wipes, pre-saturated, sterile, PROSAT® Sterile™ Low Endotoxin
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
These low endotoxin wipes are made from 100% knitted polyester and pre-saturated with a blend of 70% USP-Grade IPA and 30% water for injection (WFI).
  • Laser-sealed edges
  • Good abrasion and chemical resistance
  • Pouch can be easily resealed throughout use maintaining saturation levels
  • Each batch of wipes has a guaranteed endotoxin level of less than 1 EU/wipe
  • Validated sterile via gamma irradiation to a sterility assurance level (SAL) of 10⁻⁶
Suitable for use in the most critical pharmaceutical cleanrooms.
Suitable for use in Grade A, ISO 3More Product Information